Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172855.3 | NP_766443.2 | 1 | 537 | ||
GenBank mRNA | AK029323.1 | - | 809 | |||
BC116331.1 | - | 1 | 537 | |||
BC116332.1 | - | 1 | 537 | 40096906 |