Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCSK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004716.3 | NP_004707.2 | 7, 8 | 1554 | ||
XM_017018546.1 | XP_016874035.1 | 2, 3 | 82 | |||
GenBank mRNA | AK222907.1 | - | 7, 8 | 1094 | ||
AK312429.1 | - | 7, 8 | 1129 | |||
BC006357.2 | - | 7, 8 | 1072 | 4125594 | ||
BC010696.1 | - | 7, 8 | 1124 | 3868257 | ||
BT006870.1 | - | 5, 6 | 923 | |||
U33849.1 | - | 7, 8 | 1074 | |||
U40623.1 | - | 5, 6 | 944 |