Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004719.2 | NP_004710.2 | 15 | 4925 | ||
XM_005269230.2 | XP_005269287.2 | 15 | 4815 | |||
XM_011538984.2 | XP_011537286.1 | 16 | 6444 | |||
XM_011538985.1 | XP_011537287.1 | 15 | 4759 | |||
XM_017020217.1 | XP_016875706.1 | 16 | 4803 | |||
XM_017020219.1 | XP_016875708.1 | 16 | 5002 | |||
XM_017020220.1 | XP_016875709.1 | 15 | 4584 | |||
XM_017020221.1 | XP_016875710.1 | 14 | 5132 | |||
XM_017020222.1 | XP_016875711.1 | 13 | 5102 | |||
XM_017020223.1 | XP_016875712.1 | 16 | 5025 | |||
GenBank mRNA | AF030234.1 | - | 15 | 4873 | ||
Y11251.1 | - | 5 | 4903 |