Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYRK1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001396.3 | NP_001387.2 | 3, 4 | 410 | ||
NM_101395.2 | NP_567824.1 | 5, 6 | 584 | |||
NM_130436.2 | NP_569120.1 | 3, 4 | 1778 | |||
NM_130438.2 | NP_569122.1 | 3, 4 | 411 | |||
XM_006723976.3 | XP_006724039.1 | 4, 5 | 482 | |||
XM_006723977.3 | XP_006724040.1 | 4, 5 | 807 | |||
XM_006723978.3 | XP_006724041.1 | 4, 5 | 1536 | |||
XM_006723979.3 | XP_006724042.1 | 4, 5 | 642 | |||
XM_011529482.2 | XP_011527784.1 | 3, 4 | 504 | |||
XM_011529483.2 | XP_011527785.1 | 4, 5 | 3350 | |||
XM_011529484.2 | XP_011527786.1 | 3, 4 | 477 | |||
XM_011529485.2 | XP_011527787.1 | 3, 4 | 993 | |||
XM_017028284.1 | XP_016883773.1 | 4, 5 | 441 | |||
XM_017028285.1 | XP_016883774.1 | 4, 5 | 687 | |||
XM_017028286.1 | XP_016883775.1 | 6, 7 | 684 | |||
GenBank mRNA | AB015282.1 | - | 3, 4 | 994 | ||
AF108830.1 | - | 5, 6 | 584 | |||
D85759.1 | - | 3, 4 | 376 | |||
D86550.1 | - | 3, 4 | 1781 | |||
U52373.1 | - | 3, 4 | 410 | |||
U58496.1 | - | 3, 4 | 370 |