Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gphn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145965.2 | NP_666077.2 | 23 | 3107 | ||
NM_172952.3 | NP_766540.2 | 24 | 3098 | |||
GenBank mRNA | AK082353.1 | - | 24 | 3020 | ||
AK144569.1 | - | 23 | 3112 | |||
AK220267.1 | - | 5 | 1092 | |||
BC027112.1 | - | 12 | 1894 |