Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnft2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109902.1 | NP_001103372.1 | 11 | 1549 | ||
NM_172998.3 | NP_766586.2 | 12 | 1584 | |||
GenBank mRNA | AK046850.1 | - | 12 | 1585 | ||
AK090256.1 | - | 11 | 1645 | |||
AK148930.1 | - | 11 | 1527 | |||
BC057302.1 | - | 11 | 1511 | 6827287 |