Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173026.2 | NP_766614.2 | 4 | 1243 | ||
GenBank mRNA | AK048360.1 | - | 1 | 1524 |