Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lingo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165999.1 | NP_001159471.1 | 8 | 1512 | ||
NM_001166000.1 | NP_001159472.1 | 9 | 971 | |||
NM_001166001.1 | NP_001159473.1 | 2 | 432 | |||
NM_175516.4 | NP_780725.2 | 3 | 539 | |||
GenBank mRNA | AK035778.1 | - | 3 | 481 | ||
AK045639.1 | - | 3 | 540 | |||
AK141568.1 | - | 8 | 995 | |||
BC090619.1 | - | 9 | 899 | 6734654 | ||
BC137866.1 | - | 2 | 373 | 8860885 | ||
BC144985.1 | - | 2 | 373 | 9053533 | ||
BC145692.1 | - | 2 | 373 | 40130995 |