Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp13a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284375.1 | NP_001271304.1 | 26, 27 | 3273 | ||
NM_175650.4 | NP_783581.2 | 27, 28 | 3408 | |||
GenBank mRNA | AK083102.1 | - | 28, 29 | 3446 | ||
AK083121.1 | - | 27, 28 | 3409 | |||
AK158355.1 | - | 27, 28 | 3228 |