Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uggt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081252.2 | NP_001074721.2 | 18 | 2098 | ||
GenBank mRNA | AK046834.1 | - | 5 | 577 | ||
BC110672.1 | - | 4 | 479 |