Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hif1an | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176958.3 | NP_795932.2 | 1 | 125 | ||
GenBank mRNA | AK043636.1 | - | 1 | 111 | ||
AK084960.1 | - | 1 | 120 | |||
AK141867.1 | - | 1 | 111 | |||
AK154774.1 | - | 1 | 125 | |||
BC130013.1 | - | 1 | 129 | 40140349 | ||
BY122345.1 | - | 1 | 129 | |||
BY713373.1 | - | 1 | 111 |