Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030180.2 | NP_084456.2 | 7 | 1323 | ||
GenBank mRNA | AK036864.1 | - | 6 | 712 | ||
AK047620.1 | - | 7 | 1323 |