Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9330154K18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034078.1 | - | 3130 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_177011.1 | - | 3130 | Chr. - - [-] on Build |