Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138590.4 | NP_613056.2 | 3, 4 | 805 | ||
GenBank mRNA | AK029808.1 | - | 3, 4 | 812 | ||
AK032353.1 | - | 3, 4 | 777 | |||
AK039158.1 | - | 3, 4 | 726 | |||
AK043238.1 | - | 3, 4 | 718 | |||
AK080785.1 | - | 4, 5 | 894 | |||
AK157728.1 | - | 3, 4 | 708 | |||
BC050200.1 | - | 2, 3 | 680 | |||
BC117020.1 | - | 3, 4 | 708 | 40126339 | ||
BC117022.1 | - | 3, 4 | 708 | 40126341 | ||
BC144774.1 | - | 3, 4 | 712 | 9053322 | ||
CF732773.1 | - | 2, 3 | 396 |