Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177052.3 | NP_796026.2 | 5 | 523 | ||
GenBank mRNA | AB001434.1 | - | 2 | 182 | ||
AK043404.1 | - | 5 | 480 | |||
AK083760.1 | - | 5 | 560 |