Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177771.5 | NP_808439.2 | 10 | 3453 | ||
GenBank mRNA | AK173029.1 | - | 10 | 3453 | ||
BC025563.1 | - | 10 | 3464 | 5322893 | ||
BC034111.1 | - | 1 | 2029 | |||
BC065167.1 | - | 9 | 3149 |