Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178414.3 | NP_848501.1 | 2 | 334 | ||
GenBank mRNA | AK132127.1 | - | 2 | 335 | ||
BC048390.1 | - | 2 | 270 | 6312894 |