Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2ry6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183168.2 | NP_898991.1 | 3 | 1536 | ||
GenBank mRNA | AK142680.1 | - | 3 | 1561 | ||
AX775901.1 | - | 3 | 1496 | |||
BC027331.1 | - | 3 | 1532 | 3982042 | ||
BC064095.1 | - | 3 | 1516 | 1054566 |
Set Membership: |
G-protein coupled receptor |