Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bahcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198423.3 | NP_940815.3 | 14, 15 | 4956 | ||
GenBank mRNA | AB257855.1 | - | 13, 14 | 4958 | ||
AK147250.1 | - | 14, 15 | 4956 | |||
AK147400.1 | - | 6, 7 | 1864 | |||
BC072602.1 | - | 3, 4 | 394 | 6830770 |