Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008431.2 | NP_032457.1 | 3 | 505 | ||
GenBank mRNA | AF056492.1 | - | 3 | 543 | ||
BC119784.1 | - | 3 | 375 | 40106379 |