Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008445.2 | NP_032471.2 | 3 | 2570 | ||
GenBank mRNA | AF013116.1 | - | 3 | 2146 | ||
AK139894.1 | - | 3 | 246 | |||
AK147572.1 | - | 3 | 2372 | |||
AK220283.1 | - | 4 | 2295 | |||
BC052732.1 | - | 3 | 1727 | |||
BC053053.1 | - | 3 | 2512 | |||
BC062648.1 | - | 3 | 2245 | |||
BC127063.1 | - | 3 | 1782 | 40109965 |