Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATXN3 SIRNA Assays ›
Gene Symbol:
ATXN3
Gene Name:
ataxin 3
Gene Aliases:
AT3, ATX3, JOS, MJD, MJD1, SCA3
Chromosome Location:
Chr.14: 92058552 - 92106621 on Build GRCh38
UniGene ID:
Hs.532632
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
15507
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATXN3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127696.1
NP_001121168.1
3
334
NM_001164778.1
NP_001158250.1
4
379
NM_001164781.1
NP_001158253.1
2
169
NM_004993.5
NP_004984.2
4
379
NM_030660.4
NP_109376.1
3
214
NR_028459.1
4
379
NR_028461.1
4
379
GenBank mRNA
AB050194.1
-
3
203
AK225884.1
-
4
318
AK314919.1
-
4
331
BC033711.1
-
4
322
4393766
BC095402.1
-
4
339
30347215
BU190081.1
-
3
313
DA827537.1
-
4
379
GQ176433.1
-
4
332
GQ176434.1
-
4
331
GQ176435.1
-
4
332
GQ176440.1
-
3
167
GQ176452.1
-
3
332
GQ176454.1
-
4
332
GQ176455.1
-
3
167
GQ176462.1
-
4
332
GQ176467.1
-
3
167
GQ176486.1
-
3
287
GQ176488.1
-
4
332
GQ176489.1
-
3
287
GQ176494.1
-
4
331
GQ176498.1
-
3
167
GQ176499.1
-
4
332
GQ176504.1
-
4
332
GQ176508.1
-
4
332
GQ176510.1
-
4
332
GQ176512.1
-
4
329
GQ176516.1
-
4
332
GQ176523.1
-
4
332
GQ176526.1
-
2
122
GQ176531.1
-
4
332
GQ176532.1
-
3
332
GQ176533.1
-
4
332
GQ176536.1
-
3
167
GQ176538.1
-
4
332
GQ176539.1
-
3
167
GQ176540.1
-
3
220
GQ176548.1
-
4
332
GQ176550.1
-
4
332
GQ176551.1
-
4
331
GQ176552.1
-
4
331
GQ176560.1
-
4
332
GQ176563.1
-
4
332
GQ176565.1
-
4
332
GQ176566.1
-
3
167
GQ176568.1
-
4
332
GQ176570.1
-
4
332
GQ176573.1
-
3
167
GQ176575.1
-
4
332
GQ176576.1
-
4
332
GQ176577.1
-
4
332
GQ176578.1
-
4
332
GQ176580.1
-
4
332
GQ176581.1
-
332
GQ176589.1
-
4
332
GQ176592.1
-
3
167
GQ176593.1
-
4
331
GQ176594.1
-
4
332
GQ176597.1
-
4
332
GQ176599.1
-
4
332
GQ176602.1
-
4
350
GQ176609.1
-
332
GQ176612.1
-
4
332
GQ176615.1
-
332
GQ176616.1
-
4
332
GQ176618.1
-
158
GQ176619.1
-
4
332
GQ176621.1
-
4
332
GQ176622.1
-
4
332
GQ176624.1
-
4
332
GQ176628.1
-
332
GQ176632.1
-
4
332
GQ176636.1
-
1
153
GQ176640.1
-
4
332
GQ176642.1
-
4
332
GQ176644.1
-
4
327
GQ176645.1
-
4
332
GQ176646.1
-
4
332
GQ176658.1
-
4
332
GQ176660.1
-
3
246
GQ176665.1
-
3
167
GQ176668.1
-
4
329
GQ176671.1
-
332
GQ176673.1
-
4
332
GQ176674.1
-
4
332
GQ176675.1
-
3
167
GQ176677.1
-
4
332
GQ176679.1
-
3
167
GQ176680.1
-
4
332
GQ176684.1
-
4
332
GQ176685.1
-
4
331
GQ176686.1
-
332
GQ176687.1
-
4
332
GQ176692.1
-
4
332
GQ176693.1
-
332
GQ176694.1
-
3
167
GQ176697.1
-
4
332
GQ176698.1
-
332
GQ176699.1
-
332
GQ176700.1
-
3
167
GQ176701.1
-
4
332
GQ176703.1
-
4
332
GQ176704.1
-
4
332
GQ176708.1
-
4
332
GQ176712.1
-
3
287
GQ176714.1
-
4
332
GQ176715.1
-
3
167
GQ176719.1
-
332
GQ176721.1
-
4
332
GQ176725.1
-
4
332
GQ176728.1
-
4
332
GQ176735.1
-
4
332
GQ176737.1
-
4
332
GQ176741.1
-
332
GQ176744.1
-
3
287
GQ176745.1
-
332
GQ176746.1
-
4
332
GQ176747.1
-
332
GQ176749.1
-
4
332
GQ176750.1
-
3
167
GQ176752.1
-
332
GQ176754.1
-
3
167
GQ176756.1
-
4
332
GQ176759.1
-
4
332
GQ176763.1
-
4
332
GQ176766.1
-
4
332
GQ176771.1
-
4
332
GQ176776.1
-
332
GQ176787.1
-
4
332
GQ176791.1
-
4
332
GQ176800.1
-
332
GQ176805.1
-
4
308
GQ176806.1
-
4
331
GQ176808.1
-
332
GQ176813.1
-
3
167
GQ176814.1
-
3
166
GQ176820.1
-
4
332
GQ176822.1
-
3
167
GQ176824.1
-
4
332
GQ176825.1
-
332
GQ176826.1
-
4
332
GQ176827.1
-
4
332
GQ176829.1
-
4
332
GQ176830.1
-
303
GQ176831.1
-
332
GQ176835.1
-
3
166
GQ176836.1
-
3
167
GQ176837.1
-
4
332
GQ176838.1
-
4
332
GQ176841.1
-
4
332
GQ176846.1
-
4
332
S75313.1
-
4
345
U64820.1
-
4
367
U64821.1
-
4
330
U64822.1
-
3
307
Back To Top
More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
nucleotide-excision repair
transcription, DNA-templated
regulation of transcription, DNA-templated
ubiquitin-dependent protein catabolic process
misfolded or incompletely synthesized protein catabolic process
chemical synaptic transmission
nervous system development
regulation of cell-substrate adhesion
protein deubiquitination
actin cytoskeleton organization
cellular response to heat
monoubiquitinated protein deubiquitination
exploration behavior
proteasome-mediated ubiquitin-dependent protein catabolic process
intermediate filament cytoskeleton organization
protein K63-linked deubiquitination
histone H3 deacetylation
protein K48-linked deubiquitination
cellular response to misfolded protein
positive regulation of ERAD pathway
protein localization to cytosolic proteasome complex involved in ERAD pathway
RNA polymerase II regulatory region DNA binding
histone deacetylase activity
thiol-dependent ubiquitin-specific protease activity
protein binding
omega peptidase activity
ubiquitin protein ligase binding
identical protein binding
ATPase binding
Lys63-specific deubiquitinase activity
Lys48-specific deubiquitinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0