Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lama2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008481.2 | NP_032507.2 | 6 | 979 | ||
GenBank mRNA | AK030879.1 | - | 6 | 980 | ||
AK042011.1 | - | 6 | 1068 | |||
AK080131.1 | - | 6 | 904 | |||
S75315.1 | - | 5 | 644 | |||
U12147.1 | - | 6 | 887 |