Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008485.3 | NP_032511.3 | 7 | 1030 | ||
GenBank mRNA | L20477.1 | - | 5 | 590 | ||
U43327.2 | - | 6 | 763 |