Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161799.1 | NP_001155271.1 | 3 | 211 | ||
NM_008546.3 | NP_032572.2 | 3 | 237 | |||
GenBank mRNA | AK160213.1 | - | 3 | 213 | ||
AK164920.1 | - | 3 | 194 | |||
BC035490.1 | - | 3 | 171 | 5249342 | ||
CK389335.1 | - | 3 | 237 | |||
DV654229.1 | - | 3 | 211 | |||
L23769.1 | - | 3 | 193 |