Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008548.4 | NP_032574.1 | 5 | 1405 | ||
GenBank mRNA | AK028996.1 | - | 5 | 1320 | ||
AK031038.1 | - | 5 | 1406 | |||
AK042371.1 | - | 5 | 1341 | |||
AK043932.1 | - | 5 | 1346 | |||
AK148407.1 | - | 5 | 1346 | |||
AK154820.1 | - | 5 | 1347 | |||
AK171841.1 | - | 5 | 1348 | |||
AK172376.1 | - | 5 | 1348 | |||
AK172667.1 | - | 5 | 1347 | |||
BC015265.1 | - | 5 | 1306 | 4223319 | ||
U04299.1 | - | 4 | 1009 |