Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142169.1 | - | 8 | 1655 | ||
BC052854.1 | - | 8 | 1657 | 30058638 | ||
Z12604.1 | - | 8 | 1643 |