Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EI24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290135.1 | NP_001277064.1 | 3 | 354 | ||
NM_004879.4 | NP_004870.3 | 3 | 396 | |||
NR_110770.1 | 3 | 350 | ||||
XM_011543069.1 | XP_011541371.1 | 4 | 412 | |||
XM_011543070.1 | XP_011541372.1 | 3 | 567 | |||
XM_017018582.1 | XP_016874071.1 | 4 | 412 | |||
GenBank mRNA | AF010313.2 | - | 3 | 268 | ||
AK223029.1 | - | 3 | 325 | |||
AK291951.1 | - | 3 | 305 | |||
AK296620.1 | - | 3 | 283 | |||
AK315841.1 | - | 3 | 312 | |||
AK316539.1 | - | 3 | 260 | |||
BC002390.2 | - | 3 | 262 | 2819783 | ||
BG566671.1 | - | 3 | 370 | |||
BM450796.1 | - | 381 |