Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r15a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008654.2 | NP_032680.1 | 3 | 2266 | ||
GenBank mRNA | AK136336.1 | - | 3 | 2222 | ||
AK154697.1 | - | 2 | 2748 | |||
AK167317.1 | - | 4 | 1991 | |||
BC138485.1 | - | 3 | 2131 | 8861505 | ||
BC138486.1 | - | 3 | 2131 | 8861506 | ||
X51829.1 | - | 3 | 2211 |