Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbie | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF030896.1 | - | 2 | 392 | ||
AK008843.1 | - | 2 | 634 | |||
AK011965.1 | - | 2 | 623 | |||
AK088585.1 | - | 2 | 595 | |||
AK151450.1 | - | 2 | 631 | |||
AK151692.1 | - | 2 | 631 | |||
AK153248.1 | - | 2 | 598 | |||
BC030923.1 | - | 2 | 617 | 4909950 |