Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008740.4 | NP_032766.2 | 7 | 645 | ||
GenBank mRNA | AK042341.1 | - | 8 | 663 | ||
AK049281.1 | - | 7 | 615 | |||
AK051430.1 | - | 7 | 614 | |||
AK085086.1 | - | 7 | 599 | |||
AK141986.1 | - | 7 | 615 | |||
AK153905.1 | - | 7 | 614 | |||
AK208406.1 | - | 2 | 85 | |||
BC006627.1 | - | 7 | 650 | 3968733 | ||
BC019167.1 | - | 7 | 598 | 4506351 | ||
U10120.1 | - | 7 | 603 |