Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008740.4 | NP_032766.2 | 14 | 1661 | ||
GenBank mRNA | AK028415.1 | - | 3 | 385 | ||
AK032120.1 | - | 1 | 93 | |||
AK049281.1 | - | 14 | 1631 | |||
AK085086.1 | - | 14 | 1615 | |||
AK153905.1 | - | 14 | 1630 | |||
AK203353.1 | - | 3 | 365 | |||
AK213251.1 | - | 3 | 384 | |||
AK214505.1 | - | 3 | 303 | |||
BC006627.1 | - | 14 | 1666 | 3968733 | ||
BC019167.1 | - | 14 | 1614 | 4506351 | ||
U10120.1 | - | 14 | 1619 |