Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pet2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008821.2 | NP_032847.2 | 12 | 2097 | ||
GenBank mRNA | BC138080.1 | - | 11 | 1918 | 8861099 | |
M95564.1 | - | 12 | 2102 |