Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008830.2 | NP_032856.2 | 13 | 1589 | ||
GenBank mRNA | AK050345.1 | - | 12 | 1542 | ||
BC150687.1 | - | 13 | 1459 | 9087598 | ||
J03398.1 | - | 13 | 1589 |