Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163569.1 | NP_001157041.1 | 18 | 2138 | ||
NM_010628.3 | NP_034758.2 | 18 | 2143 | |||
GenBank mRNA | AJ132889.1 | - | 18 | 2076 | ||
AK006864.1 | - | 1 | 42 | |||
AK029506.1 | - | 18 | 2036 | |||
AK029776.1 | - | 18 | 2108 | |||
AK077169.1 | - | 10 | 1079 | |||
AK087008.1 | - | 18 | 2108 |