Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005002.4 | NP_004993.1 | 5 | 463 | ||
GenBank mRNA | AF050641.1 | - | 5 | 453 | ||
AK308588.1 | - | 5 | 444 | |||
BC003351.1 | - | 4 | 317 | |||
BC009311.2 | - | 5 | 444 | 4106931 | ||
BC015837.1 | - | 5 | 459 | 4293690 | ||
BC111546.1 | - | 5 | 435 | 5761015 | ||
CB110957.1 | - | 5 | 463 | |||
L04490.1 | - | 5 | 429 |