Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lipe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039507.2 | NP_001034596.1 | 2 | 284 | ||
NM_010719.5 | NP_034849.2 | 2 | 720 | |||
GenBank mRNA | AK019846.1 | - | 2 | 1121 | ||
AK029984.1 | - | 2 | 723 | |||
AK169858.1 | - | 2 | 279 | |||
BC021642.1 | - | 2 | 280 | 4188567 | ||
BC053502.1 | - | 2 | 344 | |||
U08188.1 | - | 1 | 805 |