Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161769.1 | NP_001155241.1 | 3, 4 | 478 | ||
NM_001161770.1 | NP_001155242.1 | 2, 3 | 610 | |||
NM_010723.3 | NP_034853.1 | 3, 4 | 765 | |||
GenBank mRNA | AF074600.1 | - | 2, 3 | 326 | ||
AF096996.1 | - | 3, 4 | 494 | |||
AF102817.1 | - | 3, 4 | 395 | |||
AK080534.1 | - | 3, 4 | 413 | |||
AK138278.1 | - | 2, 3 | 611 | |||
AK159590.1 | - | 3, 4 | 692 | |||
BC003488.1 | - | 2, 3 | 477 | 2811636 | ||
BC004661.1 | - | 3, 4 | 418 | |||
BC010278.1 | - | 3, 4 | 508 | 3589532 | ||
BC100382.1 | - | 3, 4 | 370 | 6487478 |