Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lnx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159577.1 | NP_001153049.1 | 5 | 1065 | ||
NM_001159578.1 | NP_001153050.1 | 6 | 1210 | |||
NM_001159579.1 | NP_001153051.1 | 4 | 841 | |||
NM_001159580.1 | NP_001153052.1 | 4 | 811 | |||
NM_010727.4 | NP_034857.3 | 4 | 841 | |||
GenBank mRNA | AF034745.1 | - | 5 | 1065 | ||
AF034746.1 | - | 4 | 769 | |||
AK044127.1 | - | 4 | 794 | |||
AK138326.1 | - | 4 | 812 | |||
AK161194.1 | - | 6 | 1196 | |||
BC040367.1 | - | 4 | 793 | |||
BC139771.1 | - | 4 | 769 | 40111506 | ||
KJ418422.1 | - | 4 | 539 |