Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286181.1 | NP_001273110.1 | 2 | 589 | ||
NM_001286182.1 | NP_001273111.1 | 2 | 546 | |||
NM_010728.3 | NP_034858.2 | 1 | 966 | |||
GenBank mRNA | AK135213.1 | - | 1 | 97 | ||
AK141995.1 | - | 1 | 852 | |||
AK159199.1 | - | 1 | 769 | |||
AK159241.1 | - | 1 | 745 | |||
AK159263.1 | - | 1 | 806 | |||
AK159368.1 | - | 1 | 728 | |||
AK159682.1 | - | 1 | 679 | |||
AK159796.1 | - | 1 | 740 | |||
AK160096.1 | - | 1 | 740 | |||
AK164634.1 | - | 2 | 547 | |||
AK166279.1 | - | 1 | 674 | |||
AK166386.1 | - | 810 | ||||
AK166442.1 | - | 966 | ||||
BC018439.1 | - | 1 | 817 | 2655752 | ||
M65142.1 | - | 1 | 734 |