Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286181.1 | NP_001273110.1 | 8 | 1714 | ||
NM_001286182.1 | NP_001273111.1 | 8 | 1671 | |||
NM_010728.3 | NP_034858.2 | 7 | 2091 | |||
GenBank mRNA | AK135213.1 | - | 7 | 1222 | ||
AK141995.1 | - | 7 | 1977 | |||
AK159199.1 | - | 7 | 1894 | |||
AK159241.1 | - | 7 | 1870 | |||
AK159263.1 | - | 7 | 1931 | |||
AK159368.1 | - | 7 | 1853 | |||
AK164634.1 | - | 8 | 1672 | |||
BC018439.1 | - | 7 | 1942 | 2655752 | ||
M65142.1 | - | 7 | 1859 | |||
M65143.1 | - | 4 | 717 |