Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159902.1 | NP_001153374.1 | 5 | 768 | ||
NM_010737.3 | NP_034867.3 | 6 | 867 | |||
GenBank mRNA | BC120707.1 | - | 6 | 805 | 40129630 | |
BC145045.1 | - | 5 | 706 | 9053593 | ||
M77676.1 | - | 6 | 867 | |||
M77753.1 | - | 6 | 766 |