Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mad1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010752.3 | NP_034882.2 | 14 | 1526 | ||
GenBank mRNA | AF083812.1 | - | 14 | 1533 | ||
AF083813.1 | - | 14 | 1533 | |||
AK136981.1 | - | 14 | 1529 | |||
AK169976.1 | - | 3 | 495 | |||
BC010702.1 | - | 14 | 1524 | 3156639 |