Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290229.1 | NP_001277158.1 | 3 | 372 | ||
NM_004866.5 | NP_004857.4 | 4 | 450 | |||
NR_110885.1 | 4 | 450 | ||||
XM_011543727.2 | XP_011542029.1 | 4 | 450 | |||
GenBank mRNA | AK290225.1 | - | 4 | 300 | ||
AK304452.1 | - | 3 | 264 | |||
BC009787.1 | - | 4 | 342 | |||
BC015065.1 | - | 4 | 343 | 4105068 | ||
BC034048.1 | - | 4 | 304 | 4814951 | ||
CB960371.1 | - | 450 | ||||
CR542286.1 | - | 4 | 281 | |||
DB213762.1 | - | 4 | 450 |