Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010773.2 | NP_034903.2 | 6 | 1372 | ||
GenBank mRNA | AF072243.1 | - | 6 | 1330 | ||
AK147008.1 | - | 6 | 1257 | |||
BC026698.1 | - | 6 | 1332 | |||
BC046607.1 | - | 6 | 1372 | 6399206 | ||
BC057289.1 | - | 6 | 1204 | |||
BC058864.1 | - | 6 | 1192 |