Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPDU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004870.3 | NP_004861.2 | 7 | 1096 | ||
NR_024603.1 | 7 | 1091 | ||||
XM_006721597.2 | XP_006721660.1 | 6 | 776 | |||
XM_006721598.2 | XP_006721661.1 | 6 | 1038 | |||
XM_006721599.2 | XP_006721662.1 | 6 | 837 | |||
XM_011524081.2 | XP_011522383.1 | 7 | 927 | |||
GenBank mRNA | AF038961.1 | - | 7 | 896 | ||
AF059752.1 | - | 7 | 935 | |||
AF258568.1 | - | 2 | 1065 | |||
AK055923.1 | - | 7 | 811 | |||
AK075299.1 | - | 7 | 895 | |||
AK225401.1 | - | 7 | 895 | |||
AK225432.1 | - | 7 | 891 | |||
AK300140.1 | - | 7 | 882 | |||
BC001898.1 | - | 7 | 907 | 3536216 | ||
BG776573.1 | - | 3 | 435 | |||
DQ499597.1 | - | 7 | 878 |