Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Marcksl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010807.4 | NP_034937.1 | 2 | 1145 | ||
GenBank mRNA | AK079390.1 | - | 2 | 1094 | ||
AK083913.1 | - | 2 | 1096 | |||
AK150261.1 | - | 2 | 1092 | |||
AK152990.1 | - | 2 | 1093 | |||
AK169355.1 | - | 2 | 1096 | |||
BC006757.1 | - | 2 | 1073 | 3257582 | ||
X61399.1 | - | 2 | 1082 |