Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010811.2 | NP_034941.2 | 3 | 1688 | ||
GenBank mRNA | AF074925.1 | - | 1 | 938 | ||
AK150100.1 | - | 3 | 1469 | |||
AK170298.1 | - | 3 | 1471 | |||
BC050829.1 | - | 3 | 1453 | |||
BC110479.1 | - | 1 | 1086 | 40048114 | ||
BC110480.1 | - | 1 | 1086 | 40048115 | ||
U02304.1 | - | 2 | 1450 | |||
X75885.1 | - | 1 | 1095 |