Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010833.2 | NP_034963.2 | 13 | 2165 | ||
GenBank mRNA | AK088336.1 | - | 13 | 2113 | ||
AK133819.1 | - | 13 | 2169 | |||
AK145746.1 | - | 13 | 2169 | |||
AK148721.1 | - | 3 | 668 | |||
AK149065.1 | - | 3 | 669 | |||
AK150869.1 | - | 13 | 2168 | |||
AK153210.1 | - | 13 | 2168 | |||
AK157701.1 | - | 13 | 2120 | |||
BC047366.1 | - | 13 | 2159 | 3711212 | ||
BQ712805.1 | - | 2 | 564 | |||
M86390.1 | - | 12 | 1946 | |||
S47577.1 | - | 13 | 2144 |