Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010878.2 | NP_035008.2 | 3 | 532 | ||
GenBank mRNA | AF084183.1 | - | 2 | 455 | ||
AK053155.1 | - | 329 | ||||
AK054051.1 | - | 2 | 361 | |||
AK210839.1 | - | 1 | 149 | |||
BC002015.1 | - | 3 | 532 | 3593441 |